Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ARK2181

SAFC

D-(+)-Mannose

Pharma Manufacturing

Synonym(s):

D-Mannopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
1564373
EC Number:
MDL number:
UNSPSC Code:
12352201

assay

98.0% (HPLC)

Quality Level

form

crystalline powder

impurities

Endotoxin and microbial; tested

mp

133-140 °C (lit.)

suitability

suitable for manufacturing use

SMILES string

OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m1/s1

InChI key

WQZGKKKJIJFFOK-QTVWNMPRSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brett Shannon et al.
Journal of immunology (Baltimore, Md. : 1950), 192(11), 5074-5082 (2014-04-25)
HSV-2 infection is common and generally asymptomatic, but it is associated with increased HIV susceptibility and disease progression. This may relate to herpes-mediated changes in genital and systemic immunology. Cervical cytobrushes and blood were collected from HIV-uninfected African/Caribbean women in
Charles D Murin et al.
Journal of virology, 88(17), 10177-10188 (2014-06-27)
The neutralizing anti-HIV-1 antibody 2G12 is of particular interest due to the sterilizing protection it provides from viral challenge in animal models. 2G12 is a unique, domain-exchanged antibody that binds exclusively to conserved N-linked glycans that form the high-mannose patch
Varnica Khetrapal et al.
Nucleic acids research, 43(13), e83-e83 (2015-03-25)
Creation of defined genetic mutations is a powerful method for dissecting mechanisms of bacterial disease; however, many genetic tools are only developed for laboratory strains. We have designed a modular and general negative selection strategy based on inducible toxins that
Cecilia W Huo et al.
Breast cancer research : BCR, 17, 79-79 (2015-06-05)
Mammographic density (MD), after adjustment for a women's age and body mass index, is a strong and independent risk factor for breast cancer (BC). Although the BC risk attributable to increased MD is significant in healthy women, the biological basis
Ana Sofia Carvalho et al.
Molecular & cellular proteomics : MCP, 13(12), 3294-3307 (2014-08-17)
We investigated the molecular effects of glucosamine supplements, a popular and safe alternative to nonsteroidal anti-inflammatory drugs, for decreasing pain, inflammation, and maintaining healthy joints. Numerous studies have reported an array of molecular effects after glucosamine treatment. We questioned whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service