Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4813

Supelco

3-(N-Acetylamino)-5-(N-decyl-N-methylamino)benzyl alcohol

analytical standard, for drug analysis

Synonym(s):

ADMB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34N2O2
CAS Number:
Molecular Weight:
334.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCN(C)c1cc(CO)cc(NC(C)=O)c1

InChI

1S/C20H34N2O2/c1-4-5-6-7-8-9-10-11-12-22(3)20-14-18(16-23)13-19(15-20)21-17(2)24/h13-15,23H,4-12,16H2,1-3H3,(H,21,24)

InChI key

YGFQCJGXLSFQPL-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A designed activator of protein kinase C shown to increase the surface expression of the tumor-associated antigen BCA 225 and various cellular antigens in T47D cells.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Reddy et al.
Carcinogenesis, 11(12), 2123-2126 (1990-12-01)
Repeated promotion of 7,12-dimethylbenz[alpha]anthracene-initiated cells in mouse skin with 12-O-tetradecanoylphorbol-13-acetate (TPA) induces them to grow as premalignant skin papillomas and some of these subsequently progress to carcinomas. In this study, we demonstrate that this TPA-induced progression of initiated cells to
P S Witt et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 36(2), 82-85 (2004-03-06)
New blood vessels develop from preexisting vessels in response to growth factors or hypoxic conditions. Recent studies have shown that angiopoietin 2 (ANGPT-2) plays an important role in the modulation of angiogenesis and vasculogenesis in humans and mice. The signaling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service