Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3168

Supelco

N-(4-Aminobutyl)-2-naphthalenesulfonamide hydrochloride

analytical standard, for drug analysis

Synonym(s):

W-12

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N2O2S · HCl
CAS Number:
Molecular Weight:
314.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

yellow

solubility

H2O: soluble

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].NCCCCNS(=O)(=O)c1ccc2ccccc2c1

InChI

1S/C14H18N2O2S.ClH/c15-9-3-4-10-16-19(17,18)14-8-7-12-5-1-2-6-13(12)11-14;/h1-2,5-8,11,16H,3-4,9-10,15H2;1H

InChI key

WYFVKUWCEVMLOL-UHFFFAOYSA-N

General description

Stock solutions of approx. 5 mM are suggested. Stocks should be stable for at least 6 months when stored at 4 °C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A calmodulin antagonist; inhibits Ca2+/calmodulin activated phosphodiesterase and myosin light chain kinase.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masahiko Yamagishi et al.
Cytoskeleton (Hoboken, N.J.), 77(9), 351-361 (2020-08-28)
In in vitro microtubule gliding assays, most kinesins drive the rotation of gliding microtubules around their longitudinal axes in a corkscrew motion. The corkscrewing pitch is smaller than the supertwisted protofilament pitch of microtubules, indicating that the corkscrewing pitch is
Rosa Gabriela Beltrán-López et al.
BMC evolutionary biology, 18(1), 56-56 (2018-04-22)
Volcanic and tectonic activities in conjunction with Quaternary climate are the main events that shaped the geographical distribution of genetic variation of many lineages. Poeciliopsis infans is the only poeciliid species that was able to colonize the temperate highlands of
Azusa Kage et al.
The Journal of experimental biology, 223(Pt 5) (2020-01-29)
The way the unicellular, biflagellated, green alga Chlamydomonas orients upward has long been discussed in terms of both mechanics and physiology. In this study, we focus on the mechanics, i.e. the 'passive' mechanisms, of gravitaxis. To rotate the body upwards
Ryuichi Tatsumi et al.
American journal of physiology. Cell physiology, 296(4), C922-C929 (2009-01-23)
When skeletal muscle is stretched or injured, myogenic satellite cells are activated to enter the cell cycle. This process depends on nitric oxide (NO) production by NO synthase (NOS), matrix metalloproteinase activation, release of hepatocyte growth factor (HGF) from the
H Hidaka et al.
Proceedings of the National Academy of Sciences of the United States of America, 78(7), 4354-4357 (1981-07-01)
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide (W-7) and its derivatives are putative calmodulin antagonists that bind to calmodulin and inhibit Ca2+/calmodulin-regulated enzyme activities. Autoradiographic studies using tritiated W-7 showed that this compound penetrates the cell membrane, is distributed mainly in the cytoplasm, and inhibits proliferation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service