Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2428

Supelco

Amcinonide

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H35FO7
CAS Number:
Molecular Weight:
502.57
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

[H][C@@]12C[C@@]3([H])[C@]4([H])CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]1(OC6(CCCC6)O2)C(=O)COC(C)=O

InChI

1S/C28H35FO7/c1-16(30)34-15-22(33)28-23(35-26(36-28)9-4-5-10-26)13-20-19-7-6-17-12-18(31)8-11-24(17,2)27(19,29)21(32)14-25(20,28)3/h8,11-12,19-21,23,32H,4-7,9-10,13-15H2,1-3H3/t19-,20-,21-,23+,24-,25-,27-,28+/m0/s1

InChI key

ILKJAFIWWBXGDU-MOGDOJJUSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Amcinonide is a highly potent topical corticosteroid.[1] It possesses pharmacological properties like anti-pruritic, anti-inflammatory and vasoconstrictive activity.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Cornell
Cutis, 31(5), 566-569 (1983-05-01)
Thirty-nine patients with psoriasis were treated either with amcinonide ointment 0.1 percent twice a day or with fluocinonide ointment 0.05 percent three times a day in a two-week, randomized, parallel-group, double-blind study. The severities of the skin lesions at baseline
Allergic contact dermatitis due to amcinonide.
R Hayakawa et al.
Contact dermatitis, 23(1), 49-50 (1990-07-01)
Contact dermatitis from budesonide in a nasal spray without cross-reactivity to amcinonide.
P M Gamboa et al.
Contact dermatitis, 24(3), 227-228 (1991-03-01)
E M Koch et al.
Arzneimittel-Forschung, 36(10), 1540-1544 (1986-10-01)
The results of comparative studies with corticosteroid topicals on age-associated differences are presented. In each of the test series 10 to 14 subjects were included, either young adults aged 18-24 years or older adults aged 65-76 years. Tests on vasoconstriction
Contact dermatitis due to amcinonide.
E Sasaki et al.
Contact dermatitis, 18(1), 61-62 (1988-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service