The TOC Linearity Set consists of 5 × 40 mL Glass Vials. It includes 2 × Calibration/Linearity Blanks, and 1 each of 0.250 mg/L C, 0.500 mg/L C, and 0.750mg/L C, all from NIST Sucrose.
Recommended Products
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
general analytical
personal care
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the range of the TOC Linearity Set A-2030-040?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service