Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-2025-040

Supelco

System Suitability Set - USP Source Guide 34

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.21

description

ISO 17025 & ISO Guide 34 CRM - USP TOC System Suitability Set.

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
general analytical
personal care

storage temp.

2-8°C

General description

Set includes 4 x 40 mL vials - 2 USP reagent water. 1 of 0.500 mg/L C USP Sucrose and 1 x 0.500 mg/L from USP 1,4-Benzoquinone - all CRMs are phosphoric acid stabilized - kit comes with individual certificate of analysis for each CRM

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service