Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1674

Supelco

Acemetacin

analytical standard

Synonym(s):

1-(p-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid carboxymethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18ClNO6
CAS Number:
Molecular Weight:
415.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

COc1ccc2n(c(C)c(CC(=O)OCC(O)=O)c2c1)C(=O)c3ccc(Cl)cc3

InChI

1S/C21H18ClNO6/c1-12-16(10-20(26)29-11-19(24)25)17-9-15(28-2)7-8-18(17)23(12)21(27)13-3-5-14(22)6-4-13/h3-9H,10-11H2,1-2H3,(H,24,25)

InChI key

FSQKKOOTNAMONP-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

An NSAID that specifically potentiates the cytoxicity of anthracyclines (doxorubicin, daunorubicin and epirubium). Circumvents MRP-mediated multidrug resistance.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B F Leeb et al.
Der Orthopade, 33(9), 1032-1041 (2004-05-25)
The aim of this trial was to compare acemetacin (ACE) with celecoxib (CEL) in terms of tolerability and efficacy in the treatment of osteoarthritis of the knee joint. A total of 105 patients (26-64 years old) suffering from primary osteoarthritis
Christian Dejaco et al.
Rheumatology international, 27(4), 351-355 (2006-09-08)
Dose adjustments of NSAIDs according to body weight may be reasonable for an optimal efficacy/tolerance profile in the therapy of musculoskeletal disorders, but are unusual in adults. In this post hoc analysis of a prospective, one-arm clinical study of a
Thomas Gelbrich et al.
Acta crystallographica. Section C, Crystal structure communications, 63(Pt 8), o451-o453 (2007-08-07)
The structure of the title compound (systematic name: 2-{2-[1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetoxy}acetic acid monohydrate, C(21)H(18)ClNO(6).H(2)O) contains one-dimensional infinite hydrogen-bonded chains. Molecules of acemetacin, a nonsteroidal anti-inflammatory drug, and water are linked by three independent O-H...O bonds. The central unit in each chain consists
M Gil-Flores et al.
Methods and findings in experimental and clinical pharmacology, 32(2), 101-105 (2010-04-20)
Indomethacin is a nonsteroidal anti-inflammatory drug (NSAID) used for the treatment of acute gout and inflammation. However, its use is limited due to side effects. Acemetacin is a prodrug of indomethacin that exhibits better gastric tolerability in preclinical and clinical
Y Q Hu et al.
Se pu = Chinese journal of chromatography, 17(6), 586-587 (2003-01-30)
A method of HPLC for the quantitative determination of acemetacin and indometacin in human serum is described. After being extracted with diethyl ether, acemetacin and indometacin were analyzed by reversed-phase HPLC(Spherisorb-C8) and UV-detector(254 nm), with tolbutamide as internal standard. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service