99916
Silica gel C18 on TLC Plates
chiral modified, impregnated with Cu2+ reagent, chiral-silica gel matrix, L × W 10 cm × 20 cm
About This Item
material
chiral-silica gel matrix
glass support
quality
chiral modified
impregnated with Cu2+ reagent
feature
binder Organic
fluorescent indicator
packaging
1 ea of 50 plates
manufacturer/tradename
Sigma-Aldrich
technique(s)
thin layer chromatography (TLC): suitable
L × W
10 cm × 20 cm
layer thickness
250 μm
particle size
10 μm
pore size
60 Å medium pore diameter
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service