98143
D-(−)-2,3-Butanediol
Synonym(s):
(R,R)-2,3-Butanediol
About This Item
Recommended Products
form
liquid
bp
177-179 °C (lit.)
density
0.992 g/mL at 20 °C (lit.)
storage temp.
2-8°C
SMILES string
C[C@@H](O)[C@@H](C)O
InChI
1S/C4H10O2/c1-3(5)4(2)6/h3-6H,1-2H3/t3-,4-/m1/s1
InChI key
OWBTYPJTUOEWEK-QWWZWVQMSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
185.0 °F - closed cup
flash_point_c
85 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service