Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

98042

Sigma-Aldrich

L-(−)-α-Amino-ε-caprolactam

Synonym(s):

(S)-3-Amino-hexahydro-2-azepinone, L-Lysine lactam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O
CAS Number:
Molecular Weight:
128.17
Beilstein/REAXYS Number:
4230434
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

form

crystals

storage temp.

2-8°C

SMILES string

N[C@H]1CCCCNC1=O

InChI

1S/C6H12N2O/c7-5-3-1-2-4-8-6(5)9/h5H,1-4,7H2,(H,8,9)/t5-/m0/s1

InChI key

BOWUOGIPSRVRSJ-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service