Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

97597

Supelco

7-Methyl-2H-1,5-benzodioxepin-3(4H)-one

analytical standard

Synonym(s):

7-Methylbenzo[b][1,4]dioxepin-3-one, Methylbenzodioxepinone, Watermelon ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O3
CAS Number:
Molecular Weight:
178.18
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤4.0% water (calc. from elemental analysis)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

InChI

1S/C10H10O3/c1-7-2-3-9-10(4-7)13-6-8(11)5-12-9/h2-4H,5-6H2,1H3

InChI key

SWUIQEBPZIHZQS-UHFFFAOYSA-N

General description

7-Methyl-2H-1,5-benzodioxepin-3(4H)-one, a synthetic fragrance ingredient, belongs to the group of marine odorants. It is sought for its olfactory impression of a seashore with floral notes. This benzodioxepinone possesses a molecular framework desirable for the study of structure-odor relationships (SOR).[1][2]

Application

7-Methyl-2H-1,5-benzodioxepin-3(4H)-one may be used as an analytical standard for the determination of the analyte in the olfactory evaluation of synthetic marine odorant formulations by various chromatographic techniques.[1][3][2]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure-activity relationship in the domain of odorants having marine notes
Gaudin JM, et al.
Helvetica Chimica Acta, 90(7), 1245-1265 (2007)
Synthesis of benzodioxepinone analogues via a novel synthetic route with qualitative olfactory evaluation
Drevermann B, et al.
Helvetica Chimica Acta, 90(5), 1006-1027 (2007)
Synthesis of Saturated Benzodioxepinone Analogues: Insight into the Importance of the Aromatic Ring Binding Motif for Marine Odorants
Plummer CM, et al.
European Journal of Organic Chemistry, 2015(3), 486-495 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service