Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

97593

Supelco

Steel (low alloy)

manufactured by MBH Analytical Ltd (12X 15254)

Synonym(s):

Iron-carbon alloy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

material

disks

grade

analytical standard

technique(s)

XRF: suitable

diam. × H

40 mm × 15 mm , dimensions are approximate

format

matrix material

Analysis Note

Values of analytes vary lot to lot.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service