Key Documents
97517
Sigma-Aldrich
Regulator with total pressure and outlet pressure gauge
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
General description
Pressure reducing valve with total pressure and outlet flow gauge, for flow regulation 0-2.5 L/min
Choose from one of the most recent versions:
Certificates of Analysis (COA)