About This Item
Recommended Products
grade
reference material
vapor pressure
<0.01 mmHg ( 20 °C)
assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
bp
253 °C (lit.)
mp
34-37 °C (lit.)
SMILES string
CC(C)(C)c1cccc(c1O)C(C)(C)C
InChI
1S/C14H22O/c1-13(2,3)10-8-7-9-11(12(10)15)14(4,5)6/h7-9,15H,1-6H3
InChI key
DKCPKDPYUFEZCP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
replaced by
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
227.3 °F - closed cup
flash_point_c
108.5 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service