Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96441

Supelco

Alkali metal mix for ICP

TraceCERT®, 5 elements, 100 mg/L in nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

concentration

100 mg/L in nitric acid

technique(s)

ICP: suitable

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the label.
Download your certificate at: http://www.sigma-aldrich.com.

Analysis Note

100 mg/L each: Li, Na, K, Rb and Cs in 2% nitric acid

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service