96051
2-Mercaptobenzothiazole
reference material
Synonym(s):
2-MBT, 2-Benzothiazolethiol, MBT
About This Item
Recommended Products
grade
reference material
assay
≥97.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
mp
177-181 °C (lit.)
SMILES string
Sc1nc2ccccc2s1
InChI
1S/C7H5NS2/c9-7-8-5-3-1-2-4-6(5)10-7/h1-4H,(H,8,9)
InChI key
YXIWHUQXZSMYRE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
replaced by
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
392.0 °F - closed cup
flash_point_c
200 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service