95653
Zinc (impurities)
manufactured by MBH Analytical Ltd (41X Z2)
Synonym(s):
Zinc
About This Item
material
disks
grade
analytical standard
vapor pressure
1 mmHg ( 487 °C)
resistivity
5.8 μΩ-cm, 20°C
technique(s)
XRF: suitable
diam. × H
50 mm × 20 mm , dimensions are approximate
bp
907 °C (lit.)
mp
420 °C (lit.)
density
7.133 g/mL at 25 °C (lit.)
format
matrix material
SMILES string
[Zn]
InChI
1S/Zn
InChI key
HCHKCACWOHOZIP-UHFFFAOYSA-N
Analysis Note
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
No data available
flash_point_c
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service