Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95647

Supelco

Bisulfite ionophore

Selectophore

Synonym(s):

4-[4-(Dioctylamino)-phenylazo]-3-nitro-benzaldehyde, Chromoionophore CR-514

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H42N4O3
CAS Number:
Molecular Weight:
494.67
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Quality Level

form

crystals

SMILES string

CCCCCCCCN(CCCCCCCC)c1ccc(cc1)\N=N\c2ccc(C=O)cc2[N+]([O-])=O

InChI

1S/C29H42N4O3/c1-3-5-7-9-11-13-21-32(22-14-12-10-8-6-4-2)27-18-16-26(17-19-27)30-31-28-20-15-25(24-34)23-29(28)33(35)36/h15-20,23-24H,3-14,21-22H2,1-2H3/b31-30+

InChI key

GOBXXSMKFPJVEM-NVQSTNCTSA-N

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Chromoionophore for the selective detection of HSO3 which changes color from pink to orange in the presence of bisulfite (formation of aldehyde sulfoxylate)[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerhard J Mohr
Chemical communications (Cambridge, England), (22), 2646-2647 (2003-01-04)
A new chromoreactand for the reversible optical detection of bisulfite anion is presented with a sensitivity in the range from 0.1 to 10 mM while no absorbance changes are observed for anions such as sulfate, chloride, phosphate or hydroxide.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service