Skip to Content
MilliporeSigma

95470

Xanthene

purum, ≥98.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H10O
CAS Number:
Molecular Weight:
182.22
EC Number:
202-194-4
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
133939
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥98.0% (GC)

bp

310-312 °C (lit.)

mp

101-102 °C

SMILES string

C1c2ccccc2Oc3ccccc13

InChI

1S/C13H10O/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)14-12/h1-8H,9H2

InChI key

GJCOSYZMQJWQCA-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

hcodes

pcodes

Hazard Classifications

Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library