Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95413

Supelco

Cellulose on TLC Alu foils

cellulose matrix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

material

aluminum support
cellulose matrix

feature

binder Proprietary
fluorescent indicator: no

packaging

pkg of 25 ea

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

L × W

20 cm × 20 cm

layer thickness

100 μm

particle size

2-20 μm

Looking for similar products? Visit Product Comparison Guide

General description

Cellulose, a homopolymer consisting of glucose units joined by β-1,4 bonds. These are strongly attached through inter and intramolecular hydrogen bonds and van der Waals forces resulting in microfibrils which together form fibers. They are arranged in parallel reducing ends of adjacent glucan chains.[1] Cellulose is widely used as sorbent in TLC.[2] On a cellulose coated TLC Alu foil, the substances are partitioned between the developing solvent and water coating the cellulose particles.[3]

application

Cellulose on TLC Alu foils may be used for TLC and HPTLC.

Other Notes

this product will be discontinued, please find the alternative product here :
116092

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S B Leschine
Annual review of microbiology, 49, 399-426 (1995-01-01)
In anaerobic environments rich in decaying plant material, the decomposition of cellulose is brought about by complex communities of interacting microorganisms. Because the substrate, cellulose, is insoluble, bacterial and fungal degradation occurs exocellularly, either in association with the outer cell
Jadranka Odović et al.
Iranian journal of pharmaceutical research : IJPR, 11(3), 763-770 (2012-07-01)
In this assay, the evaluation of lipophilicity of four ACE-inhibitors and hydrochlorothiazide (HCTZ) with RP-TLC on cellulose layers was described using three binary solvent systems. The selected ACE inhibitors had sufficiently different structures which can indicate the method suitability for
Joseph C. Touchstone
Practice of Thin Layer Chromatography (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service