Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95411

Supelco

Potassium phosphate buffer concentrate pH 6.8

Dissolution Media Concentrate, Dilute to 25L to conform to USP & EP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164500

form

liquid

quality

Dilute to 25L to conform to USP & EP

shelf life

limited shelf life, expiry date on the label

pH

6.78-6.82 (25 °C, after dilution)

Looking for similar products? Visit Product Comparison Guide

application

Dissolution Media Concentrates have been formulated to allow quick and easy dilution to a working solution. The label on each bottle indicates the volume of concentrate in the bottle and the volume to which it must be diluted to provide a final media solution that meets the applicable pharmacopoeia specifications. The user friendly format is designed so that each bottle is a single use unit. The instructions below illustrate just how easy it is to prepare the media.

1. Prepare a clean, dry mixing vessel of a suitable size, 6L, 10L or 25L.
2. Add purified water to approximately one third of the required volume i.e. 2L, 3L or 8L.
3. Carefully pour the entire contents of the bottle of concentrate into the vessel.
4. Rinse the bottle with purified water and add rinsing′s to the mixing vessel.
5. Add purified water to the required volume.
6. Mix thoroughly.
7. Measure the pH and proceed if within specification.
8. Adjust pH if necessary.

Preparation Note

Dilution to target volume (25 L) results in ~6.805 g/L Potassium phosphate monobasic and ~0.896 g/L Sodium hydroxide.

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service