Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95344

Supelco

(2E)-1-(2,4,4-Trimethylcyclohex-2-en-1-yl)but-2-en-1-one

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
EC Number:
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.490-1.492

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=C(/C=C/C)C1CCC(C)(C)C=C1C

InChI

1S/C13H20O/c1-5-6-12(14)11-7-8-13(3,4)9-10(11)2/h5-6,9,11H,7-8H2,1-4H3/b6-5+

InChI key

NELDPSDYTZADSA-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service