Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95313

Sigma-Aldrich

Hydrogen peroxide solution

puriss. p.a., ACS reagent, not stabilized, ≥30% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein/REAXYS Number:
3587191
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥30% (RT)
vapor pressure:
23.3 mmHg ( 30 °C)

grade

ACS reagent
puriss. p.a.

vapor pressure

23.3 mmHg ( 30 °C)

assay

≥30% (RT)

form

solution

quality

not stabilized

dilution

(for analytical testing)

impurities

≤0.002% non-volatile matter
≤0.005% free acid (as H2SO4)

density

1.11 g/mL at 20 °C

anion traces

chloride (Cl-): ≤1 mg/kg
nitrate (NO3-): ≤2 mg/kg
phosphate (PO43-): ≤2 mg/kg
sulfate (SO42-): ≤1 mg/kg

cation traces

Ag: ≤0.01 mg/kg
Al: ≤0.5 mg/kg
As: ≤0.01 mg/kg
Ba: ≤0.01 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.05 mg/kg
K: ≤0.1 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.05 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.02 mg/kg
NH4+: ≤5 mg/kg
Na: ≤2 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.01 mg/kg
Sr: ≤0.01 mg/kg
Tl: ≤0.05 mg/kg
Zn: ≤0.05 mg/kg

SMILES string

OO

storage temp.

2-8°C

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of flexible, composite, nanorod nunchucks.
Ibele ME, et al.
Journal of Materials Chemistry, 21(38), 14410-14413 (2011)
THE FLUOROMETRIC ANALYSIS OF ULTRAMICRO QUANTITIES OF HYDROGEN PEROXIDE.
A S KESTON et al.
Analytical biochemistry, 11, 1-5 (1965-04-01)
Spectrofluorimetric determination of hydrogen peroxide scavenging activity.
Pazdzioch-Czochra M and Widenska A.
Analytica Chimica Acta, 452(2), 177-184 (2002)
Shoichi Iriguchi et al.
Blood, 125(2), 370-382 (2014-10-29)
Although overexpression of T-bet, a master transcription factor in type-1 helper T lymphocytes, has been reported in several hematologic and immune diseases, its role in their pathogenesis is not fully understood. In the present study, we used transgenic model mice
Koen van der Maaden et al.
Pharmaceutical research, 31(7), 1846-1854 (2014-01-29)
The aim of the study was to develop a cheap and fast method to produce hollow microneedles and an applicator for injecting vaccines into the skin at a pre-defined depth and test the applicability of the system for dermal polio

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service