95298
Hydrogen ionophore III
Selectophore™, ≥99.0%
Synonym(s):
N,N-Dioctadecylmethylamine, Proton ionophore III
About This Item
product line
Selectophore™
Quality Level
assay
≥99.0% (NT)
≥99.0%
mp
48-49 °C
SMILES string
CCCCCCCCCCCCCCCCCCN(C)CCCCCCCCCCCCCCCCCC
InChI
1S/C37H77N/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38(3)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2/h4-37H2,1-3H3
InChI key
VFLWKHBYVIUAMP-UHFFFAOYSA-N
General description
Packaging
Other Notes
Legal Information
related product
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service