Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95297

Supelco

Hydrogen ionophore II - cocktail A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

product line

Selectophore

form

liquid

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.
package with 0.10 ml

Other Notes

H+-selective membrane solution for microelectrodes. Assay of H+-activity in single cells, extra- and intercellular liquids with microelectrodes[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F

flash_point_c

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Feng et al.
Journal of experimental botany, 71(19), 6057-6073 (2020-06-27)
Aluminum (Al) toxicity is the primary limiting factor of crop production on acid soils. Tibetan wild barley germplasm is a valuable source of potential genes for breeding barley with acid and Al tolerance. We performed microRNA and RNA sequencing using
Masaaki Suzuki et al.
Acta oto-laryngologica, 135(12), 1219-1224 (2015-09-04)
Hydrostatic positive pressure and vasoconstrictor acidified the cochlear fluids, whereas the vasodilator made the fluids alkaline. CBF might play a role in regulating cochlea fluid pH. Cochlea fluid pH is highly dependent on the HCO3(-)/CO2 buffer system. Cochlear blood flow
Honghong Wu et al.
Journal of experimental botany, 69(16), 3987-4001 (2018-06-14)
The progress in plant breeding for salinity stress tolerance is handicapped by the lack of understanding of the specificity of salt stress signalling and adaptation at the cellular and tissue levels. In this study, we used electrophysiological, fluorescence imaging, and
Kangfeng Cai et al.
International journal of molecular sciences, 20(17) (2019-08-25)
In this work, the effect of drought on K+ uptake in root and its translocation from root to shoot was investigated using six barley genotypes contrasting in drought tolerance. Results showed that drought conditions caused significant changes in K+ uptake
P Chao et al.
Pflugers Archiv : European journal of physiology, 411(2), 216-219 (1988-02-01)
A series of new neutral hydrogen ion carriers suitable for application in H+-selective microelectrodes is presented. One carrier (ETH 1907) proves to be superior to tridodecylamine currently very much in use. Microelectrodes based on ETH 1907 in an optimized membrane

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service