Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95159

Supelco

Ferrocenecarboxaldehyde

for HPLC derivatisation, LiChropur, ≥98.0% (HPLC)

Synonym(s):

Cyclopentadienyl(formylcyclopentadienyl)iron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10FeO
CAS Number:
Molecular Weight:
214.04
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0% (HPLC)

form

crystals

quality

LiChropur

mp

118-120 °C (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.[H]C(=O)[C]2[CH][CH][CH][CH]2

InChI

1S/C6H5O.C5H5.Fe/c7-5-6-3-1-2-4-6;1-2-4-5-3-1;/h1-5H;1-5H;

InChI key

UQTCQJVPLIVCAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhipeng Wang et al.
Langmuir : the ACS journal of surfaces and colloids, 27(4), 1286-1291 (2010-11-04)
Single-component microcapsules were fabricated by the in situ reaction of ferrocenecarboxaldehyde (Fc-CHO) with poly(allylamine hydrochloride) (PAH) doped inside CaCO(3) microparticles, followed by core removal. The PAH-Fc microcapsules had very thick shells with remnant PAH-Fc inside, leading to a robust capsule
John Spencer et al.
Dalton transactions (Cambridge, England : 2003), (6)(6), 918-921 (2009-01-29)
The Knoevenagel condensation of 1,3-dihydro-2H-indol-2-one with ferrocene carboxaldehyde afforded an approximate 2:1 mixture of the geometrical isomers (E)- and (Z)-3-ferrocenylmethylidene-1,3-dihydro-2H-indol-2-one respectively in an overall 67% yield; the air and solution-stable isomers were readily separated by preparative thin layer chromatography and
Mina Okochi et al.
Biotechnology and bioengineering, 90(1), 14-19 (2005-03-01)
Labeling of ferrocenecarboaldehyde (Fc-CHO) to immunoglobulin G (IgG) via formation of Schiff-base and its reduction was investigated for construction of an electrochemical probe for miniaturized amperometric flow immunoassay. Approximately eight molecules of Fc-CHO were labeled to IgG and the reversible
Kubab, N., et al.
Analusis, 17, 559-564 (1989)
C Xu et al.
Fresenius' journal of analytical chemistry, 367(6), 593-595 (2001-02-28)
A ferrocenecarboxaldehyde (FCA) labeled DNA probe is used for the first time in the study of DNA damage and protection. The electrochemically active reagent FCA was labeled successfully on to a denatured calf-thymus DNA by 1-ethyl-3- (3-dimethyl-aminopropyl) carbodiimide (EDC). The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service