Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95120

Supelco

Violuric acid monohydrate

for spectrophotometric det. of Co, ≥97.0%

Synonym(s):

2,4,5,6(1H,3H)-Pyrimidinetetrone 5-oxime, 5-Isonitrosobarbituric acid, Alloxan-5-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3N3O4 · H2O
CAS Number:
Molecular Weight:
175.10
Beilstein/REAXYS Number:
162342
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.21

Quality Level

assay

≥97.0% (T)
≥97.0%

form

solid

quality

for spectrophotometric det. of Co

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

UV/Vis spectroscopy: suitable

ign. residue

≤0.1%

mp

240-250 °C (dec.)

SMILES string

O.O\N=C1/C(=O)NC(=O)NC1=O

InChI

1S/C4H3N3O4.H2O/c8-2-1(7-11)3(9)6-4(10)5-2;/h11H,(H2,5,6,8,9,10);1H2

InChI key

YHAIHNZKUCGXRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fresenius Journal of Analytical Chemistry, 261, 45-45 (1972)
Luisaldo Sandate-Flores et al.
Plants (Basel, Switzerland), 9(11) (2020-12-04)
Mexico has a great diversity of cacti, however, many of their fruits have not been studied in greater depth. Several bioactive compounds available in cacti juices extract have demonstrated nutraceutical properties. Two cactus species are interesting for their biologically active

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service