Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94846

Supelco

Heavy metal mix I

(in 12% nitric acid), TraceCERT®, acc. to California state regulation for inhalable cannabis & cannabis products, acc. to Arkansas state regulation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

shelf life

limited shelf life, expiry date on the label

composition

As, 20 mg/L
Cd, 20 mg/L
Hg, 10 mg/L
Pb, 50 mg/L

concentration

(in 12% nitric acid)

technique(s)

ICP: suitable
gas chromatography (GC): suitable

application(s)

cannabis testing

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Suitable for cannabis testing
Suitable for ICP-OES and ICP-MS

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service