94794
Heavy metal mix II
(in 12% nitric acid), TraceCERT®, acc. to California state regulation for other cannabis & cannabis products
About This Item
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
form
liquid
shelf life
limited shelf life, expiry date on the label
composition
As, 15 mg/L
Cd, 5 mg/L
Hg, 30 mg/L
Pb, 5 mg/L
concentration
(in 12% nitric acid)
technique(s)
ICP: suitable
gas chromatography (GC): suitable
application(s)
cannabis testing
format
multi-component solution
General description
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Application
Suitable for ICP-OES and ICP-MS
Preparation Note
Legal Information
Disclaimer

signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
supp_hazards
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service