Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

94776

Supelco

Methyl hexanoate

certified reference material, TraceCERT®

Synonym(s):

Caproic acid methyl ester, Hexanoic acid methyl ester, Methyl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COOCH3
CAS Number:
Molecular Weight:
130.18
Beilstein/REAXYS Number:
1744683
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.884 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCCC(=O)OC

InChI

1S/C7H14O2/c1-3-4-5-6-7(8)9-2/h3-6H2,1-2H3

InChI key

NUKZAGXMHTUAFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Flame

signalword

Warning

hcodes

pcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

99.5 °F - closed cup

flash_point_c

37.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazutoku Ohta et al.
Journal of chromatography. A, 997(1-2), 107-116 (2003-07-02)
The application of C7 aliphatic carboxylic acids (heptanoic, 2-methylhexanoic, 5-methylhexanoic and 2,2-dimethyl-n-valeric acids) as eluents in ion-exclusion chromatography with conductimetric detection for C1-C6 aliphatic carboxylic acids (formic, acetic, propionic, isobutyric, butyric, isovaleric, valeric, isocaproic and caproic acids) was carried out
J C Knight et al.
Journal of the American Mosquito Control Association, 7(1), 37-41 (1991-03-01)
Evidence is reviewed for a dose-dependent reversal of response in mosquito oviposition, in which a compound that attracts or stimulates oviposition may repel or deter oviposition at a higher concentration. On the basis of a review of structure-activity relationships in
Teun Dekker et al.
Current biology : CB, 16(1), 101-109 (2006-01-13)
Olfaction in the fruit fly Drosophila melanogaster is increasingly understood, from ligand-receptor-neuron combinations to their axonal projection patterns into the antennal lobe . Drosophila thus offers an excellent opportunity to study the evolutionary and ecological dynamics of olfactory systems. We
David M Obenland et al.
Journal of agricultural and food chemistry, 51(11), 3367-3371 (2003-05-15)
Volatile emissions of navel orange (Citrus sinensis L. Osbeck cv. Washington) fruit were evaluated as a means for predicting and gauging freeze damage. The fruits were subjected to -5 or -7 degrees C treatments in a laboratory freezer for various
O Chukwumerije et al.
The Journal of investigative dermatology, 93(3), 349-352 (1989-09-01)
The efficacy of the methyl esters of medium chain n-alkyl fatty acids as penetration enhancers was evaluated in vitro using various animal and human skins with minoxidil as the test drug. Both methyl nonanoate and methyl caprate at a 10%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service