Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94696

Supelco

Calibrator 4,0 for FOS (Food-Oil-Sensor)

100% isopropyl myristate basis

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12164500
NACRES:
NA.21

Quality Level

assay

100% isopropyl myristate basis

form

liquid

application(s)

food and beverages
general analytical

Application

Standard for quality control of frying oils

Storage Class

10 - Combustible liquids

wgk_germany

awg

flash_point_f

>302.0 °F

flash_point_c

> 150 °C

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service