Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94678

Supelco

O-Desmethyltramadol hydrochloride

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO2· HCl
Molecular Weight:
285.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water (calc. from elemental analysis)

mp

215-221 °C

application(s)

pharmaceutical

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

CN(C)C[C@H]1CCCC[C@]1(O)C2=CC=CC(O)=C2.Cl

InChI

1S/C15H23NO2.ClH/c1-16(2)11-13-6-3-4-9-15(13,18)12-7-5-8-14(17)10-12;/h5,7-8,10,13,17-18H,3-4,6,9,11H2,1-2H3;1H/t13-,15+;/m1./s1

InChI key

IRGWVAWLHXDKIX-PBCQUBLHSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Elsi Verrinder et al.
ACS omega, 6(17), 11563-11569 (2021-06-01)
Disposable single-use electrochemical sensor strips were used for quantitative detection of small concentrations of morphine in untreated capillary whole blood. Single-walled carbon nanotube (SWCNT) networks were fabricated on a polymer substrate to produce flexible, reproducible sensor strips with integrated reference
Tania E Perez Jimenez et al.
Drug metabolism and disposition: the biological fate of chemicals, 47(1), 15-25 (2018-10-28)
Tramadol is used frequently in the management of mild to moderate pain conditions in dogs. This use is controversial because multiple reports in treated dogs demonstrate very low plasma concentrations of O-desmethyltramadol (M1), the active metabolite. The objective of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service