Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

94381

Supelco

Ginsenoside Ro

analytical standard

Synonym(s):

(3β)-28-(β-D-Glucopyranosyloxy)-28-oxoolean-12-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranosiduronic acid, Chikusetsusaponin V, Polysciasaponin P3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H76O19
CAS Number:
Molecular Weight:
957.11
UNSPSC Code:
12352200
NACRES:
NA.24
Product 94381 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O[C@@H]3C(C)(C)[C@]4([H])CC[C@@]5(C)[C@@]6(C)C([C@@](CC(C)(C)CC7)([H])[C@@]7(C(O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)=O)CC6)=CC[C@]5([H])[C@@]4(C)CC3)O[C@H](C(O)=O)[C@H]1O

InChI

1S/C48H76O19/c1-43(2)14-16-48(42(61)67-40-35(58)31(54)29(52)24(20-50)63-40)17-15-46(6)21(22(48)18-43)8-9-26-45(5)12-11-27(44(3,4)25(45)10-13-47(26,46)7)64-41-37(33(56)32(55)36(65-41)38(59)60)66-39-34(57)30(53)28(51)23(19-49)62-39/h8,22-37,39-41,49-58H,9-20H2,1-7H3,(H,59,60)/t22-,23-,24-,25?,26?,27?,28-,29-,30+,31+,32+,33+,34-,35-,36+,37-,39+,40+,41-,45+,46-,47-,48+/m1/s1

InChI key

NFZYDZXHKFHPGA-BPQKMSRTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginsenoside Ro is an oleanane-type saponin, a natural anionic biosurfactant extracted from Panax ginseng which is of high research interest due to its anti-inflammatory, anti-platelet, anticomplementary and immunomodulatory properties. It is also a health promoter and the most widely used dietary supplement.[1][2][3]

Application

Ginsenoside Ro may be used as an analytical standard for the quantification of the analyte in Panax ginseng extracts[4][5][6] by chromatography-based techniques.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Profiling and identification of the metabolites of ginsenoside Ro in rat faeces and urine after oral administration
Wang J, et al.
European Food Research and Technology, 242(2), 199-210 (2016)
T W Chan et al.
Analytical chemistry, 72(6), 1281-1287 (2000-03-31)
An LC/MS-based method is established for the differentiation and authentication of specimens and commercial samples of Panax ginseng (Oriental ginseng) and Panax quinquefolius (American ginseng). This method is based on the separation of ginsenosides present in the ginseng methanolic extracts
Hong-Ping Wang et al.
Journal of ginseng research, 40(4), 382-394 (2016-10-18)
Ginsenosides are the characteristic and principal components which manifest a variety of the biological and pharmacological activities of the roots and rhizomes of Panax ginseng (GRR). This study was carried out to qualitatively and quantitatively determine the ginsenosides in the
N Fuzzati et al.
Journal of chromatography. A, 854(1-2), 69-79 (1999-09-25)
A high-performance liquid chromatographic method was developed for electrospray mass spectrometric analysis of ginsenosides in Panax ginseng roots. The analyses were performed on a reversed-phase C18 column using a binary eluent (aqueous 8 mM NH4OAc, buffered to pH 7 with
Combination of normal-phase medium-pressure liquid chromatography and high-performance counter-current chromatography for preparation of ginsenoside-Ro from panax ginseng with high recovery and efficiency
Cheng Y, et al.
Separation and Purification Technology, 73(3), 397-402 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service