94217
Peptone Water, phosphate-buffered
suitable for microbiology, ready to use media in the bottle
Synonym(s):
Buffered Peptone Water, Tryptone Phosphate Water
About This Item
Recommended Products
Quality Level
100
200
form
liquid
shelf life
limited shelf life, expiry date on the label
composition
disodium hydrogen phosphate, 3.5 g/L
peptone, 10 g/L
potassium dihydrogen phosphate, 1.5 g/L
sodium chloride, 5 g/L
final pH
7.4±0.2 (25 °C)
application(s)
environmental
food and beverages
microbiology
storage temp.
2-8°C
suitability
Providencia spp.
nonselective for Citrobacter spp.
nonselective for Escherichia coli
nonselective for Klebsiella spp.
nonselective for Proteus spp.
nonselective for Pseudomonas spp.
nonselective for Salmonella spp.
nonselective for Shigella spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for Yersinia spp.
nonselective for bacteria (General Media)
nonselective for coliforms
nonselective for enterobacteriaceae
Looking for similar products? Visit Product Comparison Guide
Application
Packaging
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service