Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

94168

Supelco

Rhapontin

analytical standard

Synonym(s):

3,3′,5-Trihydroxy-4′-methoxystilbene 3-O-β-D-glucoside, NSC 43321, Rhaponticin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O9
CAS Number:
Molecular Weight:
420.41
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(\C=C\c2cc(O)cc(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)c2)cc1O

InChI

1S/C21H24O9/c1-28-16-5-4-11(8-15(16)24)2-3-12-6-13(23)9-14(7-12)29-21-20(27)19(26)18(25)17(10-22)30-21/h2-9,17-27H,10H2,1H3/b3-2+/t17-,18-,19+,20-,21-/m1/s1

InChI key

GKAJCVFOJGXVIA-DXKBKAGUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

LC-MS identity conformed

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service