Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94126

Supelco

2,9-Dimethyl-5-picrylamino-o-phenanthroline

p.a., suitable for suitability for complexometry, ≥90% (CHN)

Synonym(s):

2,9-Dimethyl-5-(2,4,6-trinitroanilino)-1,10-phenanthroline, N-Picryl-2,9-dimethyl-o-phenanthroline-5-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14N6O6
CAS Number:
Molecular Weight:
434.36
MDL number:
UNSPSC Code:
12161500
PubChem Substance ID:

grade

p.a.

assay

≥90% (CHN)

form

powder

technique(s)

UV/Vis spectroscopy: suitable
titration: suitable

suitability

suitable for suitability for complexometry

SMILES string

Cc1ccc2cc(Nc3c(cc(cc3[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O)c4ccc(C)nc4c2n1

InChI

1S/C20H14N6O6/c1-10-3-5-12-7-15(14-6-4-11(2)22-19(14)18(12)21-10)23-20-16(25(29)30)8-13(24(27)28)9-17(20)26(31)32/h3-9,23H,1-2H3

InChI key

YMWCRDXLDDCILP-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chromogenic reagent for the determination of lithium

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service