Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93950

Sigma-Aldrich

Ubiquitin from bovine red blood cells

≥90% (GE)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

biological source

bovine blood

assay

≥90% (GE)

form

solid

solubility

H2O: 10 mg/mL, slightly hazy, faintly yellow

UniProt accession no.

storage temp.

2-8°C

Gene Information

Other Notes

Sales restrictions may apply
Stimulation of ATP-dependent proteolysis[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K D Wilkinson et al.
The Journal of biological chemistry, 256(17), 9235-9241 (1981-09-10)
It was previously shown that ubiquitin is very similar to the polypeptide cofactor of the ATP-dependent protein degradation system from rabbit reticulocytes (Wilkinson, K. D., Urban, M. K., and Haas, A. L. (1980) J. Biol. Chem. 255, 7529-7532). We have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service