Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

93742

Supelco

Pentafluorophenylhydrazine

derivatization grade (HPLC), LiChropur, ≥98.0% (GC)

Synonym(s):

(Perfluorophenyl)hydrazine, NSC 88334, PFPH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5NHNH2
CAS Number:
Molecular Weight:
198.09
Beilstein/REAXYS Number:
747800
EC Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade (HPLC)

Quality Level

assay

≥98.0% (GC)
98.0-102.0% (wt., NT)

form

crystals

quality

LiChropur

technique(s)

HPLC: suitable

mp

74-76 °C (lit.)

SMILES string

NNc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C6H3F5N2/c7-1-2(8)4(10)6(13-12)5(11)3(1)9/h13H,12H2

InChI key

BYCUWCJUPSUFBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Pentafluorophenylhydrazine may be used as a derivatizing agent[1] in the determination of semi-volatile carbonyl compounds in air and human plasma samples, using high-resolution gas chromatography coupled with gas chromatography (HRGC-MS) and liquid chromatography atmospheric-pressure chemical-ionization mass spectrometry (LC-APCI-MS-MS), respectively.[2][1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pentafluorophenylhydrazine as a coating reagent for the HRGC-MS determination of semi-volatile carbonyl compounds in air
Cecinato A, et al.
Chromatographia, 54(3-4), 263-269 (2001)
Anže Djordjević et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 132, 110877-110877 (2020-10-24)
An unavoidable consequence of cardiac surgery is oxidative stress and resulting peroxidation of biological molecules, in particular unsaturated lipids. The detection of peroxidation metabolites in biological environments is difficult due to their large variety, small concentration, and high reactivity. One
J F Sheen et al.
Analytical and bioanalytical chemistry, 380(7-8), 891-897 (2005-02-09)
Two carbonyl compounds, nabumetone and testosterone, were derivatized with pentafluorophenyl hydrazine (PFPH) and analyzed by atmospheric-pressure chemical-ionization mass spectrometry. The PFPH derivatives underwent dissociative electron capture in negative-ion APCI (ECAPCI) and gave intense [M-20](-) ions in the mass spectra. In
Chemical derivatization to enhance ionization of anabolic steroids in LC-MS for doping-control analysis.
Athanasiadou, I., et al.
Trends in Analytical Chemistry, 42, 137-156 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service