Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93739

Supelco

Hippuric acid-(phenyl-13C6)

analytical standard

Synonym(s):

N-(Benzoyl-1,2,3,4,5,6-13C6)glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C3H9NO3
CAS Number:
Molecular Weight:
185.13
UNSPSC Code:
41116107

grade

analytical standard

isotopic purity

≥99.0%

assay

≥95.0% (HPLC)

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

SMILES string

O=C(NCC(O)=O)[13C]1=[13CH][13CH]=[13CH][13CH]=[13CH]1

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service