Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93679

Supelco

Elemental Impurities according to ICH Q3D inhalation, Standard 3

TraceCERT®, in nitric acid and hydrofluoric acid (5% nitric acid and <0.5% hydrofluoric acid ), applicable for testing acc. to USP<232>, Ph.Eur. Gen. Chapter 5.20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

Quality Level

agency

according to ICH Q3D
according to Ph. Eur.
according to USP

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

Ba, 300 mg/L
Cr, 3 mg/L
Cu, 30 mg/L
Li, 25 mg/L
Mo, 10 mg/L
Sb, 20 mg/L
Sn, 60 mg/L

concentration

in nitric acid and hydrofluoric acid (5% nitric acid and <0.5% hydrofluoric acid )

technique(s)

ICP: suitable

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST..
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Analysis Note

The concentrations are according to ICH Q3D guideline.

Other Notes

ICH Q3D metal limits changed. Replacement product is 91496

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service