Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93509

Supelco

(R)-6-Methoxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

for chiral derivatization, LiChropur, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
Beilstein/REAXYS Number:
5481541
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((chiral))
for chiral derivatization

Quality Level

assay

≥99.0% (sum of enantiomers, HPLC)
≥99.0%

form

powder

optical activity

[α]20/D +69±2°, c = 1% in ethanol

optical purity

enantiomeric ratio: ≥99.5:0.5 (HPLC)

quality

LiChropur

mp

149-155 °C

SMILES string

COc1c(C)c(C)c2O[C@](C)(CCc2c1C)C(O)=O

InChI

1S/C15H20O4/c1-8-9(2)13-11(10(3)12(8)18-5)6-7-15(4,19-13)14(16)17/h6-7H2,1-5H3,(H,16,17)/t15-/m1/s1

InChI key

MNBMVDGIDVXTOF-OAHLLOKOSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter V Cornish et al.
Molecular cell, 30(5), 578-588 (2008-06-10)
During the elongation cycle, tRNA and mRNA undergo coupled translocation through the ribosome catalyzed by elongation factor G (EF-G). Cryo-EM reconstructions of certain EF-G-containing complexes led to the proposal that the mechanism of translocation involves rotational movement between the two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service