Broad Bean Peptone is obtained by enzymatic hydrolysis of broad bean protein concentrates. This peptone demonstrates excellent growth of a wide variety of microorganisms and may serve as a substitute for meat-based peptones in certain circumstances. Broad Bean Peptone is well suited for both industrial fermentation media and in diagnostic applications where an animal-free formulation would have particular interest for security or regulatory
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Applied and environmental microbiology, 75(20), 6600-6612 (2009-08-25)
Commercial wine yeast strains of the species Saccharomyces cerevisiae have been selected to satisfy many different, and sometimes highly specific, oenological requirements. As a consequence, more than 200 different strains with significantly diverging phenotypic traits are produced globally. This genetic
Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.