Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

93483

Supelco

(+)-Cedrol

analytical standard

Synonym(s):

(1S,2R,5S,7R,8R)-2,6,6,8-Tetramethyltricyclo[5.3.1.01.5]undecan-8-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Beilstein/REAXYS Number:
2206347
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥99.0% (sum of isomers, GC)

optical activity

[α]/D +10.5±1°, c = 5 in chloroform

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

bp

273 °C (lit.)

mp

86-87 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC[C@@]3(C)O

InChI

1S/C15H26O/c1-10-5-6-11-13(2,3)12-9-15(10,11)8-7-14(12,4)16/h10-12,16H,5-9H2,1-4H3/t10-,11+,12-,14-,15+/m1/s1

InChI key

SVURIXNDRWRAFU-OGMFBOKVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(+)-Cedrol may be used as an analytical standard to investigate its repellency to little fire ants in a field bioassay.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nutrition, well-being and health (2012)
Field test for repellency of cedarwood oil and cedrol to little fire ants
Eller JF, et al.
practical laboratory medicine, 47, 71-77 (2015)
Fragrance material review on cedrol
Bhatia PS, et al.
Food And Chemical Toxicology, 46(11), S100-S102 (2008)
Nutrition, well-being and health (2012)
Samantha Dayawansa et al.
Autonomic neuroscience : basic & clinical, 108(1-2), 79-86 (2003-11-15)
It is well known that odors affect behaviors and autonomic functions. Previous studies reported that some compounds in cedar wood essence induced behavioral changes including sedative effects. In the present study, we analyzed cardiovascular and respiratory functions while subjects were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service