Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93204

Supelco

N-Decanoylglycine

analytical standard

Synonym(s):

2-(Decanoylamino)acetic acid, N-(1-Oxodecyl)glycine, Decanoylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23NO3
CAS Number:
Molecular Weight:
229.32
Beilstein/REAXYS Number:
1872050
MDL number:
UNSPSC Code:
12352200
NACRES:
NB.21

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(CNC(CCCCCCCCC)=O)=O

Biochem/physiol Actions

Decanoylglycine is an acylglycine with C-10 fatty acid group as the acyl moiety. Acylglycines possess a common amidoacetic acid moiety and are normally minor metabolites of fatty acids. Elevated levels of certain acylglycines appear in the urine and blood of patients with various fatty acid oxidation disorders. They are normally produced through the action of glycine N-acyltransferase which is an enzyme that catalyzes the chemical reaction: acyl-CoA + glycine <-> CoA + N-acylglycine.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service