Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93202

Supelco

L-Valine 2-hydroxyethyl ester p-toluenesulfonate salt

pharmaceutical impurity standard

Synonym(s):

Valaciclovir Related Compound F (USP), Valaciclovir impurity F (PhEur) p-toluenesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO3 · C7H8O3S
CAS Number:
Molecular Weight:
333.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Product 93202 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

assay

≥90.0% (TLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10% related substances (TLC)
≤5.0% solvents
≤5.0 wt. % water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)[C@H](N)C(=O)OCCO.Cc1ccc(cc1)S(O)(=O)=O

InChI

1S/C7H15NO3.C7H8O3S/c1-5(2)6(8)7(10)11-4-3-9;1-6-2-4-7(5-3-6)11(8,9)10/h5-6,9H,3-4,8H2,1-2H3;2-5H,1H3,(H,8,9,10)/t6-;/m0./s1

InChI key

KIQNQGPNCRYWKY-RGMNGODLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service