93127
Dimoxystrobin Metabolite 505M09
PESTANAL®, analytical standard
Synonym(s):
(E)-2-[(5-Hydroxycarbonyl-2-methylphenoxy)methyl]-α-methoxyimino-N-methylphenylacetamide, (E)-o-[(5-Hydroxycarbonyl-2-methylphenoxy)methyl]-2-methoxyimino-N-methylphenylacetamide
About This Item
Recommended Products
grade
analytical standard
product line
PESTANAL®
assay
≥95.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
impurities
≤1.0% water
application(s)
agriculture
format
neat
storage temp.
−20°C
General description
Application
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service