93090
Triphenylphosphine
ReagentPlus®, ≥98.5%
Synonym(s):
Phosphorustriphenyl
About This Item
Recommended Products
vapor density
9 (vs air)
vapor pressure
5 mmHg ( 20 °C)
product line
ReagentPlus®
assay
≥98.5%
reaction suitability
reagent type: ligand
impurities
~1% triphenylphosphine oxide
bp
377 °C (lit.)
mp
79-81 °C (lit.)
79-81 °C
solubility
water: soluble 0.00017 g/L at 22 °C
functional group
phosphine
SMILES string
c1ccc(cc1)P(c2ccccc2)c3ccccc3
InChI
1S/C18H15P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H
InChI key
RIOQSEWOXXDEQQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1B - STOT RE 1 Inhalation
target_organs
Central nervous system,Peripheral nervous system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 2
flash_point_f
356.0 °F - closed cup
flash_point_c
180 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service