Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93088

Supelco

4-(2-Aminoethylamino)-7-(N,N-dimethylsulfamoyl)benzofurazan

LiChropur, ≥99.0% (HPLC)

Synonym(s):

7-(2-Aminoethylamino)-N,N-dimethyl-4-benzofurazansulfonamide, DBD-ED

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5O3S
CAS Number:
Molecular Weight:
285.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((HPLC))

assay

≥99.0% (HPLC)

form

crystals

quality

LiChropur

fluorescence

λex 406 nm; λem 581 nm in 0.1 M phosphate pH 7.0

SMILES string

CN(C)S(=O)(=O)c1ccc(NCCN)c2nonc12

InChI

1S/C10H15N5O3S/c1-15(2)19(16,17)8-4-3-7(12-6-5-11)9-10(8)14-18-13-9/h3-4,12H,5-6,11H2,1-2H3

InChI key

RDMBHRPODMXKHT-UHFFFAOYSA-N

General description

DBD-ED is a fluorogenic tagging reagent mostly used in HPLC for carboxylic acids.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

New precolumn fluorescence derivatization reagent for carboxylic acids[1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Prados et al.
Analytica Chimica Acta, 344, 227-227 (1997)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service