Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93026

Supelco

3-O-Methylellagic acid

analytical standard

Synonym(s):

2,3,7-Trihydroxy-8-methoxy-[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione, 2,7,8-Trihydroxy-3-methoxychromeno[5,4,3-cde]chromene-5,10-dione, Ellagic acid 3-methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H8O8
CAS Number:
Molecular Weight:
316.22
Beilstein/REAXYS Number:
1403110
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

COc1c(O)cc2C(=O)Oc3c(O)c(O)cc4C(=O)Oc1c2-c34

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service