Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92989

Supelco

4-(Dimethylamino)benzohydrazide

for HPLC derivatization, LiChropur

Synonym(s):

4-(N,N-Dimethylamino)benzhydrazide, 4-(N,N-Dimethylamino)benzoic hydrazide, 4-(N,N-Dimethylamino)benzoylhydrazine, 4-(N,NDimethylamino)benzoic acid hydrazide, p(Nme2)-Ph-CONHNH2, NSC 219987

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O
CAS Number:
Molecular Weight:
179.22
Beilstein/REAXYS Number:
2210856
MDL number:
UNSPSC Code:
41121800
PubChem Substance ID:
NACRES:
NB.21

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

assay

≥99.0% (HPLC)

form

powder

quality

LiChropur

SMILES string

O=C(NN)C1=CC=C(N(C)C)C=C1

InChI

1S/C9H13N3O/c1-12(2)8-5-3-7(4-6-8)9(13)11-10/h3-6H,10H2,1-2H3,(H,11,13)

InChI key

HITIGLAGJBMISF-UHFFFAOYSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service